4-Cyanophenylhydrazine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Hydrazinylbenzonitrile hydrochloride
A118245-100G
A118245-10G
A118245-25G
A118245-500G
A118245-1G
A118245-5G
103799-014EA
5
USD
103799-014
103824-902
77136-364
77136-366
77136-368
77136-370
77311-692
77311-694
4-Cyanophenylhydrazine hydrochloride 98%
4-Cyanophenylhydrazine hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₈ClN₃ Boiling Pt: 325.9 °C (760 mm Hg) Melting Pt: 241…244 °C Flash Pt: 150.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00673994 CAS Number: 2863-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...