4H-Pyran-4-one 95%

Supplier: Ambeed
Warning

Synonyms: gamma-Pyrone

A120439-10G A120439-250MG A120439-1G
103799-934EA 66 USD
103799-934 77149-168 77149-170 77297-764
4H-Pyran-4-one 95%
4H-Pyran-4-one

Precaución: for Research Use Only

Formula: C₅H₄O₂
MW: 96.09 g/mol
Boiling Pt: 212.5 °C (760 mm Hg)
Melting Pt: 32…34 °C
Flash Pt: 101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006576
CAS Number: 108-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR