4-Hexylaniline 95%

Supplier: Ambeed
Warning

A184131-1G A184131-5G A184131-100G A184131-25G A184131-10G
103815-716EA 11 USD
103815-716 103846-276 77083-892 77083-894 77287-358 77301-882 77748-195
4-Hexylaniline 95%
4-Hexylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₉N
MW: 177.29 g/mol
Boiling Pt: 283.8 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007927
CAS Number: 33228-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR