4-Hexen-1-ol 97%

Supplier: Ambeed
Warning

A239119-100G A239119-10G A239119-1G A239119-25G A239119-5G
103811-268EA 14 USD
103811-268 103851-066 77288-696 77288-698 77304-140 77304-142 77304-144
4-Hexen-1-ol 97%
4-Hexen-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₂O
Boiling Pt: 158.5 °C (760 mm Hg)
Flash Pt: 61.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00009713
CAS Number: 6126-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR