Chromone 97%

Supplier: Ambeed
Warning

Synonyms: 4H-Chromen-4-one

A113610-100G A113610-25G
103798-254EA 11 USD
103798-254 103824-094 77610-908 77685-007
Chromone 97%
Chromone

Precaución: Keep in a dark place, sealed in dry, room temperature

Formula: C₉H₆O₂
MW: 146.14 g/mol
Boiling Pt: 239 °C (760 mm Hg)
Melting Pt: 55…60 °C
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00024064
CAS Number: 491-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR