4-Formylbenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Terephthalaldehydic acid, 4-Carboxybenzaldehyde

A112685-100G A112685-10G A112685-25G A112685-500G A112685-5G
103808-098EA 165 USD
103808-098 103823-914 77233-656 77233-658 77233-660 77233-662 77297-198
4-Formylbenzoic acid 98%
4-Formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆O₃
MW: 150.13 g/mol
Boiling Pt: 332.6 °C (760 mm Hg)
Melting Pt: 247 °C
Flash Pt: 169.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00006951
CAS Number: 619-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR