4-Cyano-2-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-Formyl-3-methoxybenzonitrile

A206573-1G A206573-25G A206573-5G A206573-10G A206573-250MG
103817-714EA 10 USD
103817-714 103848-390 77086-650 77086-652 77086-654 77260-932 77260-934
4-Cyano-2-methoxybenzaldehyde 98%
4-Cyano-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.157 g/mol
Boiling Pt: 318.2 °C (760 mm Hg)
Melting Pt: 109…110 °C
Flash Pt: 138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07779491
CAS Number: 21962-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR