4-Formyl-3-hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

A115469-100MG A115469-250MG
103798-568EA 107 USD
103798-568 103824-424 77160-974 77891-826
4-Formyl-3-hydroxybenzoic acid 98%
4-Formyl-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆O₄
Boiling Pt: 355 °C (760 mm Hg)
Melting Pt: 235…240 °C
Flash Pt: 182.7 °C
Storage Temperature: Ambient
MDL Number: MFCD05865188
CAS Number: 619-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR