4-Formyl-2-methylthiazole 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-1,3-thiazole-4-carboxaldehyde, 2-Methylthiazole-4-carbaldehyde

A373891-1G A373891-25G A373891-5G A373891-10G A373891-250MG
103826-544EA 11 USD
103826-544 103837-562 76840-834 76840-836 76840-838 76989-906 76989-908
4-Formyl-2-methylthiazole 95%
4-Formyl-2-methylthiazole

Precaución: for Research Use Only

Formula: C₅H₅NOS
MW: 127.1643 g/mol
Boiling Pt: 219 °C (760 mm Hg)
Melting Pt: 56…58 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06655197
CAS Number: 20949-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR