4-Fluoropyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

A148314-100MG A148314-250MG A148314-1G A148314-5G
103803-914EA 37 USD
103803-914 103842-028 77259-118 77259-120 77317-478 77317-480
4-Fluoropyridin-2(1H)-one 97%
4-Fluoropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₄FNO
Boiling Pt: 251.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07437952
CAS Number: 96530-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR