4-Fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A107392-1000G A107392-10G A107392-25G A107392-500G A107392-5G A107392-100G
103822-954EA 5 USD
103822-954 77283-584 77283-586 77283-588 77283-590 77283-592 77549-068
4-Fluorophenylboronic acid 98%
4-Fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BFO₂
Melting Pt: 262…265 °C
Flash Pt: 110.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00039136
CAS Number: 1765-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR