(4-Fluorophenyl)acetone 97%
Supplier: Ambeed
|
Warning
|
A142166-100G
A142166-10G
A142166-1G
A142166-25G
A142166-5G
103803-112EA
9
USD
103803-112
77285-796
77285-798
77316-326
77316-328
77316-330
(4-Fluorophenyl)acetone 97%
(4-Fluorophenyl)acetone
Precaución: for Research Use Only
Formula:
C₉H₉FO Boiling Pt: 218.1 °C (760 mm Hg) Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000362 CAS Number: 459-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...