4-Fluoro-N-methyl-3-nitrobenzamide 95%

Supplier: Ambeed
Warning

A163457-100MG A163457-250MG A163457-1G
103805-730EA 27 USD
103805-730 103843-990 77150-996 77150-998 77320-088
4-Fluoro-N-methyl-3-nitrobenzamide 95%
4-Fluoro-N-methyl-3-nitrobenzamide

Precaución: for Research Use Only

Formula: C₈H₇FN₂O₃
MW: 198.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11855197
CAS Number: 475216-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR