4-Fluoro-N-isopropylaniline 96%

Supplier: Ambeed
Warning

Synonyms: N-Isopropyl-4-fluoroaniline

A137104-250MG A137104-5G A137104-1G
103799-360EA 10 USD
103799-360 103802-406 77285-574 77298-886 77315-360
4-Fluoro-N-isopropylaniline 96%
4-Fluoro-N-isopropylaniline

Precaución: for Research Use Only

Formula: C₉H₁₂FN
Boiling Pt: 213.9 °C (760 mm Hg)
Flash Pt: 83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00270760
CAS Number: 70441-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR