N,N-Dimethyl 4-fluorobenzenesulfonamide 95%

Supplier: Ambeed
Warning

A190952-250MG A190952-1G
103816-336EA 46 USD
103816-336 103846-942 76873-054 77084-772
N,N-Dimethyl 4-fluorobenzenesulfonamide 95%
N,N-Dimethyl 4-fluorobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₈H₁₀FNO₂S
Boiling Pt: 279.6±42.0 °C (760 mm Hg)
Flash Pt: 122.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01211990
CAS Number: 383-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR