4-Fluoroindole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-1H-indole

A135552-100G A135552-10G A135552-5G A135552-1G A135552-25G
103802-176EA 6 USD
103802-176 76767-444 76767-446 77298-796 77315-014 77315-016
4-Fluoroindole 98%
4-Fluoroindole

Precaución: for Research Use Only

Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 258 °C (760 mm Hg)
Melting Pt: 30…32 °C
Storage Temperature: Freezer
MDL Number: MFCD00055992
CAS Number: 387-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR