4-Fluorocinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2E)-3-(4-fluorophenyl)prop-2-enoic acid, p-Fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, trans-4-Fluorocinnamic acid

A203867-100G A203867-10G A203867-500G A203867-25G A203867-5G
103848-160EA 6 USD
103848-160 76691-280 76691-282 76691-284 77086-300 77302-878
4-Fluorocinnamic acid 98%
4-Fluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₂
MW: 166.15 g/mol
Boiling Pt: 281.6 °C (760 mm Hg)
Melting Pt: 207…210 °C
Flash Pt: 124.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00004395
CAS Number: 459-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR