4-Fluorobenzoic acid 97%

Supplier: Ambeed
Warning

A201998-1000G A201998-100G A201998-25G A201998-500G
103847-982EA 5 USD
103847-982 77139-350 77139-352 77139-354 77685-510
4-Fluorobenzoic acid 97%
4-Fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 253.7 °C (760 mm Hg)
Melting Pt: 184 °C
Flash Pt: 107.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00002530
CAS Number: 456-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR