4-Fluorobenzo[d]thiazol-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-fluorobenzothiazole

A188527-1G A188527-25G A188527-5G A188527-10G
103816-112EA 11 USD
103816-112 103846-708 77084-458 77084-460 77084-462 77248-486
4-Fluorobenzo[d]thiazol-2-amine 97%
4-Fluorobenzo[d]thiazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₅FN₂S
MW: 168.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04448824
CAS Number: 20358-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR