4-Fluorobenzamidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Fluorophenylformamidine hydrochloride

A202378-1G A202378-250MG A202378-25G
103817-378EA 18 USD
103817-378 103848-018 77086-130 77164-628 77302-808
4-Fluorobenzamidine hydrochloride 97%
4-Fluorobenzamidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈ClFN₂
MW: 174.61
Boiling Pt: 243.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04114421
CAS Number: 456-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR