4-Fluoro-7-azaindole 97%

Supplier: Ambeed
Warning

A112427-100MG A112427-250MG A112427-1G
103798-010EA 113 USD
103798-010 103823-844 77284-244 77284-246 77310-648
4-Fluoro-7-azaindole 97%
4-Fluoro-7-azaindole

Precaución: for Research Use Only

Formula: C₇H₅FN₂
MW: 136.1300 g/mol
Melting Pt: 118…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08272231
CAS Number: 640735-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR