4-Fluoro-5-iodopyridin-2-amine 95%

Supplier: Ambeed
Warning

A146149-100MG A146149-250MG
103803-634EA 27 USD
103803-634 103841-720 77010-534 77010-536
4-Fluoro-5-iodopyridin-2-amine 95%
4-Fluoro-5-iodopyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄FIN₂
MW: 238 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27922772
CAS Number: 1708974-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR