4-Fluoro-5-hydroxy-2-methylindole 98%

Supplier: Ambeed
Warning

A118397-10G A118397-250MG A118397-5G
103799-050EA 18 USD
103799-050 77094-338 77094-340 77297-596
4-Fluoro-5-hydroxy-2-methylindole 98%
4-Fluoro-5-hydroxy-2-methylindole

Precaución: for Research Use Only

Formula: C₉H₈FNO
MW: 165.1600 g/mol
Boiling Pt: 327.3 °C (760 mm Hg)
Flash Pt: 151.761 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368140
CAS Number: 288385-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR