4-Fluoro-3-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A108457-100G A108457-10G A108457-25G A108457-1G A108457-5G A108457-500G
103797-348EA 5 USD
103797-348 103823-128 77008-786 77008-788 77008-790 77309-966 77309-968 77682-994
4-Fluoro-3-nitrobenzonitrile 98%
4-Fluoro-3-nitrobenzonitrile

Precaución: For research use only.

Formula: C₇H₃FN₂O₂
Boiling Pt: 470.9 °C (760 mm Hg)
Flash Pt: 121.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01632197
CAS Number: 1009-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR