4-Fluoro-3-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A115196-100G A115196-10G A115196-25G A115196-500G A115196-5G
103824-376EA 5 USD
103824-376 77271-178 77271-180 77271-182 77271-184 77311-192
4-Fluoro-3-nitrobenzoic acid 98%
4-Fluoro-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FNO₄
MW: 185.11 g/mol
Boiling Pt: 356.9 °C (760 mm Hg)
Melting Pt: 120…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00007058
CAS Number: 453-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR