4-Fluoro-3-nitrobenzamide 98%

Supplier: Ambeed
Warning

A832110-100MG A832110-250MG A832110-5G
103841-172EA 164 USD
103841-172 76832-488 76832-490 76845-406
4-Fluoro-3-nitrobenzamide 98%
4-Fluoro-3-nitrobenzamide

Precaución: for Research Use Only

Formula: C₇H₅FN₂O₃
MW: 184.13 g/mol
Boiling Pt: 293.313 °C (760 mm Hg)
Flash Pt: 131.192 °C
Storage Temperature: Ambient
MDL Number: MFCD11110229
CAS Number: 349-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR