4-Fluoro-3-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A138437-100G A138437-10G A138437-500G A138437-1G A138437-25G A138437-5G
103802-614EA 6 USD
103802-614 76972-462 76972-464 76972-466 77298-992 77298-994 77315-644
4-Fluoro-3-nitrobenzaldehyde 98%
4-Fluoro-3-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄FNO₃
Boiling Pt: 286 °C (760 mm Hg)
Melting Pt: 44…47 °C
Flash Pt: 126.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01861388
CAS Number: 42564-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR