4-Fluoro-m-cresol 97%

Supplier: Ambeed
Warning

A109494-100G A109494-10G A109494-1G A109494-25G A109494-5G A109494-500G
103823-310EA 14 USD
103823-310 76765-720 76765-722 76765-724 76765-726 77283-864 77623-906
4-Fluoro-m-cresol 97%
4-Fluoro-m-cresol

Precaución: for Research Use Only

Formula: C₇H₇FO
MW: 126.13 g/mol
Boiling Pt: 205.8 °C (760 mm Hg)
Melting Pt: 32 °C
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00042296
CAS Number: 452-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR