4-Fluoro-3-methoxyphenol 97%

Supplier: Ambeed
Warning

A309519-1G A309519-25G A309519-100G
103814-590EA 20 USD
103814-590 103835-770 76838-534 76838-536 76875-750
4-Fluoro-3-methoxyphenol 97%
4-Fluoro-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇FO₂
Boiling Pt: 253.8 °C (760 mm Hg)
Flash Pt: 123.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10698649
CAS Number: 117902-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR