4-Fluoro-m-anisidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-3-anisidine, 4-Fluoro-3-methoxyaniline, 5-Amino-2-fluoroanisole

A230823-100G A230823-10G A230823-500G A230823-5G A230823-1G A230823-25G
103810-730EA 9 USD
103810-730 103850-476 76771-136 76771-138 76771-140 77089-348 77303-886 77303-888
4-Fluoro-m-anisidine 98%
4-Fluoro-m-anisidine

Precaución: for Research Use Only

Formula: C₇H₈FNO
Boiling Pt: 252.5 °C (760 mm Hg)
Melting Pt: 66…68 °C
Flash Pt: 106.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00665789
CAS Number: 64465-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR