4-Fluoro-3-iodobenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A107225-250MG
A107225-1G
A107225-10G
A107225-100G
A107225-25G
103797-156EA
11
USD
103797-156
103822-922
77025-506
77309-772
77666-560
77871-418
77871-419
4-Fluoro-3-iodobenzoic acid 95%
4-Fluoro-3-iodobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄FIO₂ Melting Pt: 173…175 °C Flash Pt: 159 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03788556 CAS Number: 403-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...