4-Fluoro-3-hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

A135253-100G A135253-10G A135253-250MG A135253-1G A135253-25G A135253-5G
103802-120EA 6 USD
103802-120 76767-426 76767-428 76767-430 77298-764 77314-918 77314-920
4-Fluoro-3-hydroxybenzoic acid 98%
4-Fluoro-3-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅FO₃
MW: 156.11 g/mol
Boiling Pt: 324 °C (760 mm Hg)
Melting Pt: 214…218 °C
Flash Pt: 149.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00269641
CAS Number: 51446-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR