3-Cyano-4-fluorobenzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-formylbenzonitrile

A215639-25G A215639-5G A215639-100G A215639-10G
103849-212EA 14 USD
103849-212 77087-752 77087-754 77192-768 77192-770
3-Cyano-4-fluorobenzaldehyde 97%
3-Cyano-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄FNO
MW: 149.1219 g/mol
Boiling Pt: 215.6 °C (760 mm Hg)
Melting Pt: 80…84 °C
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD01863558
CAS Number: 218301-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR