α,α,α,4-Tetrafluoro-m-toluic acid 98%

Supplier: Ambeed
Warning

A472138-100G A472138-10G A472138-500G A472138-1G A472138-25G A472138-5G
103828-040EA 5 USD
103828-040 103839-114 76774-282 76774-284 76774-286 76843-028 76843-030 76843-032
α,α,α,4-Tetrafluoro-m-toluic acid 98%
α,α,α,4-Tetrafluoro-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
Boiling Pt: 256.9±40.0 °C (760 mm Hg)
Melting Pt: 114…116 °C
Flash Pt: 109.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00061294
CAS Number: 67515-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR