5-Amino-2-fluorobenzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-FLUORO-3-(4,4,5,5-TETRAMETHYL-1,3,2-DIOXABOROLAN-2-YL)ANILINE, 5-AMINO-2-FLUOROPHENYLBORONIC ACID PINACOL ESTER, 5-Amino-2-fluorophenylboronic acid pinacol ester

A103708-100MG A103708-250MG A103708-1G A103708-5G
103796-602EA 117 USD
103796-602 103822-312 77160-340 77160-342 77309-180 77309-182
5-Amino-2-fluorobenzeneboronic acid pinacol ester 97%
5-Amino-2-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₂
Boiling Pt: 356.4±32.0 °C (760 mm Hg)
Flash Pt: 169.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16996293
CAS Number: 1152441-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR