4-Fluoro-2-nitropyridine 97%

Supplier: Ambeed
Warning

A252447-1G A252447-5G A252447-100MG A252447-250MG
103812-044EA 127 USD
103812-044 103851-888 77091-302 77091-304 77113-484 77113-486
4-Fluoro-2-nitropyridine 97%
4-Fluoro-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃FN₂O₂
MW: 142.09 g/mol
Boiling Pt: 261.4 °C (760 mm Hg)
Flash Pt: 111.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08277259
CAS Number: 884495-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR