4-Fluoro-2-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A383838-25G A383838-5G A383838-250MG A383838-1G
103826-752EA 7 USD
103826-752 103837-788 76841-128 76841-130 77168-094 77306-950
4-Fluoro-2-nitrobenzaldehyde 98%
4-Fluoro-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄FNO₃
Boiling Pt: 294.2 °C (760 mm Hg)
Flash Pt: 131.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01666439
CAS Number: 2923-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR