4-Fluoro-2-methyl-6-nitrophenol 98%

Supplier: Ambeed
Danger

A101873-1G A101873-250MG
103796-186EA 29 USD
103796-186 77008-346 77282-854 77889-706
4-Fluoro-2-methyl-6-nitrophenol 98%
4-Fluoro-2-methyl-6-nitrophenol

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
Boiling Pt: 253.6±35.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD27923653
CAS Number: 1588441-30-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR