4-Fluoro-2-methoxypyridine 98%

Supplier: Ambeed
Warning

A340090-250MG A340090-1G A340090-25G A340090-10G
103825-804EA 16 USD
103825-804 103836-778 76724-328 76839-792 77306-522 77593-982
4-Fluoro-2-methoxypyridine 98%
4-Fluoro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆FNO
Boiling Pt: 131.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09835267
CAS Number: 96530-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR