4-Fluoro-2-methoxyphenol 98%

Supplier: Ambeed
Warning

A443689-1G A443689-100G A443689-250MG A443689-25G A443689-5G A443689-10G
103827-700EA 13 USD
103827-700 76842-548 77143-406 77143-408 77143-410 77143-412 77746-905
4-Fluoro-2-methoxyphenol 98%
4-Fluoro-2-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇FO₂
Boiling Pt: 195 °C (760 mm Hg)
Flash Pt: 101.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00070797
CAS Number: 450-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR