4-Fluoro-o-anisaldehyde 98%

Supplier: Ambeed
Warning

A320303-1G A320303-25G A320303-5G A320303-100G A320303-10G A320303-250MG
103825-230EA 14 USD
103825-230 103836-164 76839-032 76839-034 76839-036 76978-060 76978-062 76978-064
4-Fluoro-o-anisaldehyde 98%
4-Fluoro-o-anisaldehyde

Precaución: For research use only

Formula: C₈H₇FO₂
Boiling Pt: 224.6 °C (760 mm Hg)
Melting Pt: 55 °C
Flash Pt: 87.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00143318
CAS Number: 450-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR