4-Fluoro-2-methoxyaniline 98%

Supplier: Ambeed
Warning

A123691-100G A123691-25G A123691-1G A123691-5G
103800-430EA 5 USD
103800-430 76781-870 76781-872 77312-546 77312-548
4-Fluoro-2-methoxyaniline 98%
4-Fluoro-2-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₈FNO
Boiling Pt: 207.9 °C (760 mm Hg)
Flash Pt: 79.53 °C
Storage Temperature: Ambient
MDL Number: MFCD00077536
CAS Number: 450-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR