4-Fluoro-2-iodophenol 98%

Supplier: Ambeed
Warning

A144967-10G A144967-250MG A144967-1G A144967-5G
103803-472EA 9 USD
103803-472 103841-550 77027-830 77027-832 77316-812 77316-814
4-Fluoro-2-iodophenol 98%
4-Fluoro-2-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄FIO
MW: 237.998 g/mol
Boiling Pt: 196 °C (760 mm Hg)
Melting Pt: 120…123 °C
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD15527539
CAS Number: 2713-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR