2-Cyano-5-fluorobenzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-2-formylbenzonitrile

A156473-250MG A156473-1G A156473-5G
103804-888EA 55 USD
103804-888 103843-090 76783-612 77318-872 77318-874
2-Cyano-5-fluorobenzaldehyde 97%
2-Cyano-5-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄FNO
MW: 149.122 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16619724
CAS Number: 894779-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR