4-Fluoro-2-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-1-methoxy-2-nitrobenzene

A124557-100G A124557-10G A124557-25G A124557-5G
76766-816EA 71 USD
76766-816 76766-818 76766-820 77298-002
4-Fluoro-2-nitroanisole 98%
4-Fluoro-2-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆FNO₃
Boiling Pt: 272.4 °C (760 mm Hg)
Melting Pt: 62…64 °C
Flash Pt: 118.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013375
CAS Number: 445-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR