4-Fluoro-1H-pyrazole 95%

Supplier: Ambeed
Danger

A262895-1G A262895-5G A262895-100MG A262895-10G A262895-250MG A262895-25G
103812-586EA 9 USD
103812-586 103852-482 77092-112 77092-114 77098-844 77098-846 77098-848 77550-648
4-Fluoro-1H-pyrazole 95%
4-Fluoro-1H-pyrazole

Precaución: for Research Use Only

Formula: C₃H₃FN₂
Boiling Pt: 192.7 °C (760 mm Hg)
Melting Pt: 38…40 °C
Flash Pt: 70.3 °C
Storage Temperature: Ambient
MDL Number: MFCD10699125
CAS Number: 35277-02-2
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR