4-Fluoro-1H-indole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A100680-10G A100680-250MG A100680-1G A100680-5G
103795-570EA 23 USD
103795-570 103821-266 76970-138 76970-140 77296-800 77296-802
4-Fluoro-1H-indole-2-carboxylic acid 98%
4-Fluoro-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆FNO₂
MW: 179.1500 g/mol
Boiling Pt: 422.2 °C (760 mm Hg)
Flash Pt: 209.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02664471
CAS Number: 399-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR