4-Fluoro-1,3-dioxolan-2-one 98%

Supplier: Ambeed
Warning

Synonyms: Fluoroethylene Carbonate, 4-Fluoro-2-oxo-1,3-dioxolane

A135675-100G A135675-10G A135675-500G A135675-25G
103808-514EA 175 USD
103808-514 76798-038 76798-040 76798-042 77298-808
4-Fluoro-1,3-dioxolan-2-one 98%
4-Fluoro-1,3-dioxolan-2-one

Precaución: for Research Use Only

Formula: C₃H₃FO₃
MW: 106.05 g/mol
Boiling Pt: 249.5 °C (760 mm Hg)
Melting Pt: 18…23 °C
Flash Pt: 102.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06247543
CAS Number: 114435-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR