4-Ethynylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Pyridylacetylene

A241293-1G A241293-5G A241293-10G A241293-100MG
103811-404EA 43 USD
103811-404 103851-208 77090-372 77090-374 77288-736 77568-342
4-Ethynylpyridine 97%
4-Ethynylpyridine

Precaución: For research use only.

Formula: C₇H₅N
MW: 103.12 g/mol
Melting Pt: 98 °C
Storage Temperature: Freezer
MDL Number: MFCD04038420
CAS Number: 2510-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR