4-Ethynylpyridin-2-amine 96%

Supplier: Ambeed
Warning

A159003-100MG A159003-1G
103805-210EA 195 USD
103805-210 103843-422 77235-780 77319-284
4-Ethynylpyridin-2-amine 96%
4-Ethynylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₇H₆N₂
MW: 118.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13189750
CAS Number: 1094679-27-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR